25(OH) Vitamin D ELISA kit
Lieferant: ENZO LIFE SCIENCES
Rapid, easy-to-use ELISA kit to quantify 25(OH) Vitamin D for bone metabolism, nutrition, endocrinology, immunity and cancer.
For the quantitative determination of human 25(OH) Vitamin D in plasma and serum.
The 25(OH) Vitamin D ELISA kit is a complete, colorimetric, competitive immunoassay kit for the quantitative determination of 25(OH) Vitamin D in plasma and serum with results in just 1.5 hours.
Bestellinformation: Contains Microtiter Plate, Conjugate Concentrate, Antibody, Dissociation Buffer, Conjugate Diluent, Sample Diluent, Wash Buffer Concentrate, Standards, Substrate, Stop Solution, Assay Layout.