TXB2 ELISA Kits
Lieferant: ENZO LIFE SCIENCES
ADI901002
ENZOADI901002EA 2180 EUR
ENZOADI901002
TXB2 ELISA Kits
Tests ELISAs
For the quantitative determination of TXB2 in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample types include cell lysate, coronary effluent, liver perfusate, platelets, rectal dialysate, and whole blood.
The TXB2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 in a convenient 96 well plate based assay. This kit is highly cited in peer-reviewed publications in a diverse assortment of sample types including mucosa, tissue, serum, plasma, urine, and much more. The TXB2 EIA kit provides easy-to-use stable liquid color-coded reagents that reduce errors.
Bestellinformation: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-054 (1x96 wells).