6-keto-PGF1α ELISA Kits
Lieferant: ENZO LIFE SCIENCES
ADI901004
ENZOADI901004EA 2030 EUR
ENZOADI901004
6-keto-PGF1α ELISA Kits
Tests ELISAs
For the quantitative determination of 6-keto-PGF1α in culture supernatants, serum, saliva, and urine from any species.
The 6-keto-PGF1α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 1α EIA kit has been fully validated in-house in a wide variety of sample matrices.
Bestellinformation: Contains DxS Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-056 (1x96 wells).