Chemiluminescent labelling kit
Lieferant: ENZO LIFE SCIENCES
ADI907001
ENZOADI907001EA 659 EUR
ENZOADI907001
Chemiluminescent labelling kit
Proteinkonjugate und Markierungen
A complete kit for the labeling of antibodies, proteins, peptides, and nucleic acids with Acridinium NHS Ester.
Chemiluminescent acridinium esters are commonly used to label antibodies, proteins, nucleic acids, and peptides containing primary amine groups. These chemiluminescent molecules are frequently for use as detection reagents in immunoassays as a safe alternative to radioactive tracers. The Chemiluminescent labeling kit provides the necessary components for quick, efficient, and easy acridinium labeling.
Bestellinformation: Contains Acridinium NHS Ester, Dry DMF, Bicarbonate labeling buffer, 10% Lysine, Gel filtration column, Column buffer concentrate, Set of Trigger solutions.