Hydrogen peroxide chemiluminescent detection Kit
Lieferant: ENZO LIFE SCIENCES
ADI907012
ENZOADI907012EA 469 EUR
ENZOADI907012
Hydrogen peroxide chemiluminescent detection Kit
Tests Zelltests
For the quantitative determination of hydrogen peroxide in culture supernatants and cell lysates from any species.
  • Immediate results in less than 10 minutes
  • Extremely sensitive quantitation of hydrogen peroxide
The Hydrogen Peroxide chemiluminescent detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide. This kit offers more sensitivity over the Hydrogen Peroxide colorimetric detection kit but is not recommended for samples that contain phenol red or hemoglobin.
Bestellinformation: Contains White Microtiter plate, Standard, Substrate, Trigger solution.