Hydrogen peroxide colorimetric detection kit
Lieferant: ENZO LIFE SCIENCES
ADI907015
ENZOADI907015EA 444 EUR
ENZOADI907015
Hydrogen peroxide colorimetric detection kit
Tests Zelltests
Hydrogen peroxide colorimetric detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide in a wide variety of samples.