Hydrogen peroxide fluorometric detection kit
Lieferant: ENZO LIFE SCIENCES
ADI907028
ENZOADI907028EA 542 EUR
ENZOADI907028
Hydrogen peroxide fluorometric detection kit
Tests Zelltests
The Hydrogen Peroxide Fluorometric Detection Kit provides fast results in just 10 minutes for the measurement of either hydrogen peroxide or peroxidase activity in a convenient one-step homogeneous assay.
  • Flexible in measure hydrogen peroxide or peroxidase activity
  • Higher throughput analysis
  • Fast results
For the measurement of hydrogen peroxide in cell lysates and culture supernatants from any species.
Lieferumfang: Reaction buffer concentrate, Detection Reagent, Hydrogen peroxide, Horseradish peroxidase