PGE2 high sensitivity ELISA kit, plasma
Lieferant: ENZO LIFE SCIENCES
ADI930001
ENZOADI930001EA 539 EUR
ENZOADI930001
PGE2 high sensitivity ELISA kit, plasma
Tests ELISAs
Ultrasensitive PGE2 ELISA kit for inflammation and eicosanoid research.
For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species. Cited sample type includes plasma.
The PGE2 high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour.
Bestellinformation: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-766 (5x96 wells).