PGE2 high sensitivity ELISA kit
Lieferant: ENZO LIFE SCIENCES
ADI931001
ENZOADI931001EA 2150 EUR
ENZOADI931001
PGE2 high sensitivity ELISA kit
Tests ELISAs
Ultrasensitive PGE2 ELISA kit for inflammation and eicosanoid research.
For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species.
The PGE2 high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour.
Bestellinformation: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-762 (1x96 wells).