IHC PBS tablets
Lieferant: ENZO LIFE SCIENCES
ADI9502400100
ENZOADI9502400100EA 165 EUR
ENZOADI9502400100
IHC PBS tablets
Puffer Assay-Puffer
Solid. 100 ml PBS (1X) tablets containing 137 mM sodium chloride, 2.7 mM potassium chloride, and 10 mM phosphate buffer.