IHC-Verdünner (primäre AK)
Lieferant: ENZO LIFE SCIENCES
Immunohistochemistry diluent (primary Ab) can be used as a diluent to prepare primary antibodies and negative controls.
- Optimal pH with stabilising proteins in the IHC diluent (primary Ab)
- Maximum stability of primary antibodies
- Good penetration of primary antibodies via selected concentrations of sodium chloride and detergents
Specifically designed to stabilise primary antibodies and reduce non-specific background staining observed during immunostaining.