IHC Fluoromount, HIGHDEF®
Lieferant: ENZO LIFE SCIENCES
ADI9502600025
ENZOADI9502600025EA
185
EUR
ENZOADI9502600025
IHC Fluoromount, HIGHDEF®
Färbemittel
Mikroskopie- und Färbereagenzien
Fluoromount is an aqueous-based permanent, high stability mounting medium designed for fluorogenic labelling and ideal for long-term storage of processed tissue.
- High definition visualisation for IHC, IF, ISH, FISH
- It is compatible with FITC, phycoerythrin, phycocyanin, and allophycocyanin
- Can also be used with other applications
Formulation: Liquid, mounting medium containing <0,1% sodium azide.