ImmunoSet® HSP27 high sensitivity (human), ELISA development set
Lieferant: ENZO LIFE SCIENCES
ADI960076
ENZOADI960076EA 714 EUR
ENZOADI960076
ImmunoSet® HSP27 high sensitivity (human), ELISA development set
Tests ELISAs
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of human HSP27 in cell lysates, plasma, and serum.
The ImmunoSet® HSP27 high sensitivity (human), EIA development set contains sufficient reagents for the development of 5×96 well EIA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own EIA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules.
Bestellinformation: Contains Capture antibody, Standard, Detection antibody, SA-HRP.