DNA damage ELISA Kit
Lieferant: ENZO LIFE SCIENCES
Rapid DNA damage ELISA kit for cancer, apoptosis and oxidative stress.
For the measurement of DNA Damage (8-OHdG) in serum, saliva, and urine from any species. Cited sample types include plasma, seminal fluids, cell lysates, culture supernatants and DNA extracts.
The DNA Damage EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 2,5 hours. This kit has been validated in-house in a variety of sample matrices and is supplied in a convenient 96 well plate format. Protocol (xls) available upon request.
Bestellinformation: Contains Microtiter plate, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.