HSP60 (human), ELISA Kit
Lieferant: ENZO LIFE SCIENCES
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of HSP60 in cell lysates, serum, and tissue of human origin.
The HSP60 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Bestellinformation: Contains Microtiter plate, 5X Extraction reagent, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, HRP Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.