HO-1 (human), ELISA Kit
Lieferant: ENZO LIFE SCIENCES
ADIEKS800
ENZOADIEKS800EA 910 EUR
ENZOADIEKS800
HO-1 (human), ELISA Kit
Tests ELISAs
Highly sensitive HO-1 ELISA kit enabling detection of HO-1 in cell lysates and tissue extracts in < 2,5 hours.
For the quantitative determination of human heme oxygenase-1 in cell lysates and tissue extracts. Cited sample types include serum and plasma.
The HO-1 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 2,5 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Bestellinformation: Contains Microtiter plate, 5X Extraction reagent, Standard, Sample diluent, Wash buffer concentrate, Antibody, Antibody diluent, HRP Conjugate, Conjugate diluent, TMB Substrate, Stop solution 2.