HO-1 (rat), ELISA Kit
Lieferant: ENZO LIFE SCIENCES
Highly sensitive colorimetric immunometric enzyme immunoassay kit with results in < 3 hours.
For the quantitative determination of rat HO-1 in cell lysates, plasma, serum, and tissue. Cited sample type includes bronchoalveolar lavage fluid.
The HO-1 (rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Bestellinformation: Contains Microtiter plate, 5X Extraction reagent 2, Standard, Sample diluent, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2.