Recombinant thioredoxin (from E. coli), HIS tag

Lieferant: ENZO LIFE SCIENCES

ADISPP8921
ENZOADISPP8921EA 833 EUR
ENZOADISPP8921
Recombinant thioredoxin (from E. coli), HIS tag
Proteine und Peptide

Thioredoxin (Trx) is a redox protein of approximately 12 kDa. Its primary domain is conserved across a number of Trx family members and contains a conserved catalytic site Cys-Gly-Pro-Cys. It is ubiquitous and found in many organisms from bacteria to mammals. Trx has been shown to function in cell proliferation, redox signaling and inhibition of apoptosis.


  • Produced from E. coli
  • E. coli thioredoxin is fused at the N-terminus to a His-tag
  • Purified by multi-step chromatography
  • Supplied in 50 mM TRIS, pH 7,5, containing 100 mM sodium chloride and 1 mM EDTA

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR