N-Methyl-D-asparaginsäure ≥98% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-Methyl-D-asparaginic Acid, NMDA, Me-D-Asp-OH, (R)-2-(Methylamino)succinic acid

ALX105009M050
ENZOALX105009M050EA 66 EUR
ENZOALX105009M050
N-Methyl-D-asparaginsäure ≥98% (durch TLC)
N-Methyl-D-asparaginsäure

NMDA receptor agonist.


Selective ionotropic NMDA receptor agonist. Excitatory amino acid.


Soluble in dilute aqueous base; slightly soluble in water or 100% ethanol.


Bitte beachten Research use only.

Formel: C₅H₉NO₄
Molecular Weight: 147,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004226
CAS-Nummer: 6384-92-5
Merck Index: 14,06662
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c7c24c425586bc9de1bcb27b17a93d3455e26e8&vid=0af38c9958670b08e47ca44adab9245958aa8960&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity ≥98% (TLC)
Identity Determined by NMR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR