S-Tritylcystein
Lieferant: ENZO LIFE SCIENCES
Synonyme:
H-Cys(Trt)-OH
ALX105011M500
ENZOALX105011M500EA
66
EUR
ENZOALX105011M500
S-Tritylcystein
S-Trityl-L-cysteine
Cell cycle blocker
Potent, cell permeable cysteine thioether displaying anti-mitotic and anti-tumor properties. Inhibits mitosis by blocking basal and microtubule-activated ATPase activities of the mitotic kinesin Eg5 (IC50=1µM and 140nm). Induces mitotic arrest in HeLa cells (IC50=700nM) and inhibits the growth of NCI tumor cell line panel (GI50=1.31µM).
Formel:
C₂₂H₂₁NO₂S Molecular Weight: 363,5 g/mol |
MDL:
MFCD00002611 CAS-Nummer: 2799-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...