D-NMMA monoacetate ≥99%
Lieferant: ENZO LIFE SCIENCES
Synonyme:
NG-Monomethyl-D-arginine . monoacetate
ALX106002M025
ENZOALX106002M025EA
186
EUR
ENZOALX106002M025
D-NMMA monoacetate ≥99%
D-NMMA monoacetate
NOS inhibitor
D-NMMA is inactive as an inhibitor of NOS and may be used to explore non-specific effects of L-NMMA.
Formel:
C₇H₁₆N₄O₂·CH₃COOH Molecular Weight: 248,3 g/mol |
CAS-Nummer:
137694-75-8 |
|
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...