NG,NG'-Dimethyl-L-arginine dihydrochloride ≥98% (durch TLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
SDMA dihydrochloride, SDMA.2HCl, Symmetric dimethylarginine dihydrochloride, H-Arg(di-Me)-OH.2HCl (symmetrical)
ALX106008M025
ENZOALX106008M025EA
165
EUR
ENZOALX106008M025
NG,NG'-Dimethyl-L-arginine dihydrochloride ≥98% (durch TLC)
NG,NG'-Dimethyl-L-arginine dihydrochloride
Does not inhibit nitric oxide synthesis, both in vitro and in vivo.
Does not inhibit nitric oxide synthesis, both in vitro and in vivo.
Hygroscopic and soluble in water (100 mg/ml).
Bitte beachten Research use only.
Formel:
C₈H₁₈N₄O₂·2HCl Molecular Weight: 275,2 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
30344-00-4 |
Spezifikation Testergebnisse
Purity | ≥98% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...