L-Cycloserin
Lieferant: ENZO LIFE SCIENCES
Synonyme:
L-4-Aminoisoxazolidin-3-on, (S)-(-)-Cycloserin
ALX106037M100
ENZOALX106037M100EA
162
EUR
ENZOALX106037M100
L-Cycloserin
L-Cycloserin
Formel:
C₃H₆N₂O₂ Molecular Weight: 102.09 g/mol |
MDL:
MFCD00064324 CAS-Nummer: 339-72-0 EINECS: 206-427-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...