L-Theanin ≥98% (durch titrimetrische Analyse)
Lieferant: ENZO LIFE SCIENCES
|
Achtung
|
Synonyme:
(S)-2-Amino-5-(ethylamino)-5-oxopentanoic acid, L-gamma-Glutamylethylamide, N-gamma-Ethyl-L-glutamine
Glutamate receptor ligand.
Amino acid analog of glutamine, uniquely found in green tea. Increases brain serotonin, dopamine and GABA levels and binds to AMPA, kainate and NMDA receptors. Has been shown to exert neuroprotective effects in animal models possibly through its antagonistic effects on group 1 metabotrophic glutamate receptors. Behavioural studies with rats suggest L-theanine to positiveley affect learning and memory processes. Promotes self-renewal of human embryonic stem cells.
Soluble in water (50 mg/ml).
Formel:
C₇H₁₄N₂O₃ Molecular Weight: 174,20 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00059653 CAS-Nummer: 3081-61-6 EINECS: 221-379-0 |
Spezifikation Testergebnisse
Purity | ≥98% (Titration) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...