Blocking peptide for Adiponectin receptor 1 pAb Blocking Peptide
Lieferant: ENZO LIFE SCIENCES
ALX151045C100
ENZOALX151045C100EA 288 EUR
ENZOALX151045C100
Blocking peptide for Adiponectin receptor 1 pAb Blocking Peptide
Proteine und Peptide
Synthetic peptide corresponding to aa 345-369 of human, mouse or rat adiponectin receptor 1 (AdipoR1).