ETA receptor antagonist [BQ-610]
Lieferant: ENZO LIFE SCIENCES
ALX155011P001
ENZOALX155011P001EA 237 EUR
ENZOALX155011P001
ETA receptor antagonist [BQ-610]
Proteine und Peptide
30-fold more potent ETA antagonist than BQ-123 in pig aortic smooth muscle membrane assay, same activity as BQ-123 when tested on pig cerebellum membrane.
Bestellinformation: Quantity: *Sold as net peptide weight.