Human EGF receptor (652-657)
Lieferant: ENZO LIFE SCIENCES
ALX155023M001
ENZOALX155023M001EA 80.3 EUR
ENZOALX155023M001
Human EGF receptor (652-657)
Proteine und Peptide
Specific PKC substrate
Specific substrate (aa 652-657 of mature EGF receptor, without signal peptide (aa 1-24)) for protein kinase C (PKC).