Synthetic melittin
Lieferant: ENZO LIFE SCIENCES
ALX162007M001
ALX162007MC05
ENZOALX162007M001EA
306
EUR
ENZOALX162007M001
ENZOALX162007MC05
Synthetic melittin
Proteine und Peptide
PLA2 stimulatory bee venom peptide. Sequence is similar to Phospholipase A2 Activating Peptide (Prod. No. ALX-165-007). Activates PLA2 (5-fold at 10 µg/ml) in BC3H1 cells. For additional technical information see melittin (natural) (Prod. No. ALX-162-006).
- PLA2 activator