NBD peptide
Lieferant: ENZO LIFE SCIENCES
ALX1630111
ENZOALX1630111EA
468
EUR
ENZOALX1630111
NBD peptide
Proteine und Peptide
For increased cell permeability the peptide was covalently linked to the 10 aa recognized by the TAT transporter.
Bestellinformation: Quantity: 50 µl