Human recombinant Arginase 1
Lieferant: ENZO LIFE SCIENCES
ALX201081C020
ENZOALX201081C020EA 457 EUR
ENZOALX201081C020
Human recombinant Arginase 1
Enzyme
Produced in E. coli. Full length human arginase I.