Human recombinant SUMO2 (1-93) (from E. coli)
Lieferant: ENZO LIFE SCIENCES
ALX201089C500
ENZOALX201089C500EA 414 EUR
ENZOALX201089C500
Human recombinant SUMO2 (1-93) (from E. coli)
Proteine und Peptide
Active form of SUMO-2
Produced in E. coli.
Active form of human SUMO-2 for conjugation to protein substrates.