Human recombinant KillerTRAIL™ (soluble) (from E. coli)
Lieferant: ENZO LIFE SCIENCES
ALX201123C500
ENZOALX201123C500EA
2160
EUR
ENZOALX201123C500
Human recombinant KillerTRAIL™ (soluble) (from E. coli)
Proteine und Peptide
Produced in E. coli. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a His-tag and a linker peptide.