Human recombinant Matriptase 2
Lieferant: ENZO LIFE SCIENCES
ALX2017521
ENZOALX2017521EA
514
EUR
ENZOALX2017521
Human recombinant Matriptase 2
Enzyme
Produced in Drosophila Schneider 2 (S2) cells. Recombinant human matriptase-2 (aa 78-811) is fused at the C-terminus to a V5-His-tag.
- Trypsin-like serine protease.
Matriptase-2 is a potent negative regulator of hepcidin.