Human recombinant Activin-A
Lieferant: ENZO LIFE SCIENCES
ALX2018090010 ALX2018090100
ENZOALX2018090010EA 348 EUR
ENZOALX2018090010 ENZOALX2018090100
Human recombinant Activin-A
Proteine und Peptide
Produced in E. coli.
  • Carrier-free
Non-glycosylated homodimer, containing two 117 amino acid chains.