Human recombinant Activin-A
Lieferant: ENZO LIFE SCIENCES
ALX2018090010
ALX2018090100
ENZOALX2018090010EA
348
EUR
ENZOALX2018090010
ENZOALX2018090100
Human recombinant Activin-A
Proteine und Peptide
Produced in E. coli.
- Carrier-free
Non-glycosylated homodimer, containing two 117 amino acid chains.