Human recombinant IL-6
Lieferant: ENZO LIFE SCIENCES
ALX2018180020
ENZOALX2018180020EA
310
EUR
ENZOALX2018180020
Human recombinant IL-6
Proteine und Peptide
Produced in E. coli. Non-glycosylated protein, containing 184 amino acids.
- Carrier-free