Human recombinant eNOS
Lieferant: ENZO LIFE SCIENCES
ALX2018530010
ENZOALX2018530010EA 576 EUR
ENZOALX2018530010
Human recombinant eNOS
Proteine und Peptide
Produced in Sf9 cells. Untagged human eNOS.