Poly(ADP-ribose), standard
Lieferant: ENZO LIFE SCIENCES
ALX202043C001
ENZOALX202043C001EA 516 EUR
ENZOALX202043C001
Poly(ADP-ribose), standard
Proteine und Peptide
Released from automodified PARP-1 by base treatment and purified by boronate affinity.
Bestellinformation: Quantity: 1µg representing 1.8 nanomoles of 2-300 units of ADP-ribose.