CA-074
Lieferant: ENZO LIFE SCIENCES
ALX260017M001
ENZOALX260017M001EA 479 EUR
ENZOALX260017M001
CA-074
Proteine und Peptide
Cathepsin B inhibitor
Potent and specific inhibitor of cathepsin B in vitro and in vivo. Free acid form used in vitro or in isolated enzyme studies but not in whole cells. Can discriminate between cathepsin B and L/S forms.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7831ac4e9dde6c2ded063271aeb2bad1eea556d3&vid=22351cc971f1c96457a3a6e2337be18e950a435f&prid=web&clid=SAPDAM