CA-074
Lieferant: ENZO LIFE SCIENCES
ALX260017M001
ENZOALX260017M001EA
479
EUR
ENZOALX260017M001
CA-074
Proteine und Peptide
Cathepsin B inhibitor
Potent and specific inhibitor of cathepsin B in vitro and in vivo. Free acid form used in vitro or in isolated enzyme studies but not in whole cells. Can discriminate between cathepsin B and L/S forms.