Mca-YVADAPK(Dnp)-OH
Lieferant: ENZO LIFE SCIENCES
ALX260023M001
ENZOALX260023M001EA 523 EUR
ENZOALX260023M001
Mca-YVADAPK(Dnp)-OH
Proteine und Peptide
Substrate for caspase-1 and -4
Specific, highly fluorescent substrate for caspase-1. Also substrate for caspase-4.