Ac-VDVAD-AMC
Lieferant: ENZO LIFE SCIENCES
ALX260060M005
ENZOALX260060M005EA
299
EUR
ENZOALX260060M005
Ac-VDVAD-AMC
Proteine und Peptide
Caspase-2 inhibitor
Fluorogenic substrate for caspase-2. Preferred cleavage site for caspase-2 (Km=80 μM).