N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone ≥95% (durch TLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: Boc-Asp(OMe)-fluoromethyl ketone, Methyl (S)-3-((tert-butoxycarbonyl)amino)-5-fluoro-4-oxopentanoate

ALX260071M001 ALX260071M005
ENZOALX260071M001EA 438 EUR
ENZOALX260071M001 ENZOALX260071M005
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone ≥95% (durch TLC)
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone

Irreversible, cell permeable broad-spectrum caspase inhibitor.


  • Caspase inhibitor


White to off-white powder.


Soluble in DMSO or dimethyl formamide.

Formel: C₁₁H₁₈FNO₅
Molecular Weight: 263.265 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03453073
CAS-Nummer: 187389-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR