Z-YVAD(OMe)-FMK
Lieferant: ENZO LIFE SCIENCES
ALX260074M001
ENZOALX260074M001EA 569 EUR
ENZOALX260074M001
Z-YVAD(OMe)-FMK
Proteine und Peptide
Inhibitor of caspase-1 and -4
Irreversible, cell permeable inhibitor of caspase-1. Inhibits also caspase-4.