Ac-LEHD-CHO
Lieferant: ENZO LIFE SCIENCES
ALX260079M005
ENZOALX260079M005EA
465
EUR
ENZOALX260079M005
Ac-LEHD-CHO
Proteine und Peptide
Caspase-9 inhibitor
Reversible aldehyde inhibitor of caspase-9 and related cysteine proteases. Sequence is from combinatorial experiments.