Biotinyl-VAD(OMe)-FMK
Lieferant: ENZO LIFE SCIENCES
ALX260098M001
ENZOALX260098M001EA
876
EUR
ENZOALX260098M001
Biotinyl-VAD(OMe)-FMK
Proteine und Peptide
Caspase inhibitor
Irreversible, cell permeable broad-spectrum caspase inhibitor.