Dnp-Pro-Leu-Gly-Met-Trp-Ser-Arg-OH
Lieferant: ENZO LIFE SCIENCES
ALX260120M005
ENZOALX260120M005EA
335
EUR
ENZOALX260120M005
Dnp-Pro-Leu-Gly-Met-Trp-Ser-Arg-OH
Proteine und Peptide
Substrate for MMP-2 and -9
Fluorogenic substrate for MMP-2 and MMP-9. Excitation: 280 nm; emission 360 nm.