Mca-Pro-Leu-Ala-Nva-Dap(Dnp)-Ala-Arg-NH2
Lieferant: ENZO LIFE SCIENCES
ALX260123M001
ENZOALX260123M001EA
369
EUR
ENZOALX260123M001
Mca-Pro-Leu-Ala-Nva-Dap(Dnp)-Ala-Arg-NH2
Proteine und Peptide
MMP-2 substrate
Fluorogenic substrate for MMP-2.