Z-Phe-Arg-pNA
Lieferant: ENZO LIFE SCIENCES
ALX260130M005
ENZOALX260130M005EA 56.1 EUR
ENZOALX260130M005
Z-Phe-Arg-pNA
Proteine und Peptide
Cathepsin substrate
Chromogenic substrate for Cathepsins B, K, L, and S, papain, trypsin, and plasma kallikrein. This substrate is likely to be cleaved by other cathepsins (see Z-Phe-Arg-AMC). Release of pNA is monitored at 405 to 410 nm (pNA ε410=8800 M-1cm-1). Useful for inhibitor screening and kinetic analysis.