MMP-3 inhibitor ≥95% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
Synonyme:
Stromelysin-1 inhibitor, Ac-RCGVPD-NH2, Matrix metalloproteinase 3 inhibitor
ALX260165M005
ENZOALX260165M005EA
215
EUR
ENZOALX260165M005
MMP-3 inhibitor ≥95% (durch HPLC)
MMP-3 inhibitor
Inhibitor of matrix metalloproteinase 3 (MMP-3) (IC50=5µM).
White to off-white solid.
Soluble in water.
Formel:
C₂₇H₄₆N₁₀O₉S Molecular Weight: 686,8 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
158841-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...