Tyrphostin AG 1478 ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: N-(3-Chlorophenyl)-6,7-dimethoxy-4-quinazolinamine, N-(3-Chlorophenyl)-6,7-dimethoxyquinazolin-4-amine

ALX270036M005
ENZOALX270036M005EA 159 EUR
ENZOALX270036M005
Tyrphostin AG 1478 ≥98% (durch HPLC)
Tyrphostin AG 1478

EGFR inhibitor


Potent and selective inhibitor of EGFR (IC50 = 3nM). Reduces EGF-stimulated DNA synthesis in Rat-1 fibroblasts by ~75% at 0.25μM. Blocks EGF-dependent src-family kinase activation and p21/Cip 1/WAF1 induction in A431 cells. Also inhibits H2O2-induced stimulation of amphiregulin expression in rat gastric epithelium (RGM 1) cells.

Formel: C₁₆H₁₄ClN₃O₂
Molecular Weight: 315.76 g/mol
MDL: MFCD00270914
CAS-Nummer: 153436-53-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR