Aristolochic acid ≥97%

Lieferant: ENZO LIFE SCIENCES

ALX270047M025
ENZOALX270047M025EA 72.6 EUR
ENZOALX270047M025
Aristolochic acid ≥97%
Aristolochic acid

Reversible noncompetitive PLA2 inhibitor.


Phospholipase A2 (PLA2) inhibitor active against the enzymes found in many snake venoms as well as those of human platelets and synovial fluids. Inhibits ionophore-stimulated PLA2 activity in human neutrophils. Has been shown to be nephropathic and carcinogenic.


Isolated from Aristolochia clematis.

Formel: C₁₇H₁₁NO₇
Molecular Weight: 341.275 g/mol
MDL: MFCD00238539
CAS-Nummer: 313-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR